Assay Details
| Gene Symbol: LOC4327816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757213.2 | XP_015612699.1 | 11 - 12 | 3883 | 67 | |
| GenBank mRNA | AK122066.1 | - | 11 - 12 | 3880 | 67 | |
More Information
Set Membership: |
|

LOC4327816
Genomic Map