Assay Details
| Gene Symbol: LOC4327816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757213.2 | XP_015612699.1 | 12 - 13 | 4943 | 139 | |
| GenBank mRNA | AK060263.1 | - | 1 - 2 | 944 | 139 | |
| AK122066.1 | - | 12 - 13 | 4940 | 139 | ||
More Information
Set Membership: |
|

LOC4327816
Genomic Map