Assay Details
| Gene Symbol: LOC4327816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757213.2 | XP_015612699.1 | 5 - 6 | 604 | 122 | |
| GenBank mRNA | AK099871.1 | - | 5 - 6 | 610 | 122 | |
| AK122066.1 | - | 5 - 6 | 601 | 122 | ||
More Information
Set Membership: |
|

LOC4327816
Genomic Map