Assay Details
| Gene Symbol: LOC4327816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757213.2 | XP_015612699.1 | 9 - 10 | 2384 | 77 | |
| GenBank mRNA | AK099871.1 | - | 9 - 10 | 3087 | 77 | |
| AK122066.1 | - | 9 - 10 | 2381 | 77 | ||
More Information
Set Membership: |
|

LOC4327816
Genomic Map