Assay Details
| Gene Symbol: LOC4328088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772149.2 | XP_015627635.1 | 1 - 2 | 290 | 69 | |
| GenBank mRNA | AK061815.1 | - | 1 - 2 | 186 | 69 | |
| AK102022.1 | - | 1 - 2 | 184 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772149.2 transcripts.
Set Membership: |
|

LOC4328088
Genomic Map