Assay Details
| Gene Symbol: LOC4328135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768101.2 | XP_015623587.1 | 6 - 7 | 1040 | 80 | |
| XM_015768102.2 | 7 - 8 | 900 | 80 | |||
| GenBank mRNA | AK064363.1 | - | 6 - 7 | 1041 | 80 | |
| AK101344.1 | - | 7 - 8 | 895 | 80 | ||
| X60375.1 | - | 7 - 8 | 822 | 80 | ||
More Information
Set Membership: |
|

LOC4328135
Genomic Map