Assay Details
| Gene Symbol: LOC4328196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771786.2 | XP_015627272.1 | 3 - 4 | 402 | 131 | |
| GenBank mRNA | AK066913.1 | - | 3 - 4 | 366 | 131 | |
| KF737321.1 | - | 3 - 4 | 372 | 131 | ||
| KY170871.1 | - | 3 - 4 | 281 | 131 | ||
More Information
Set Membership: |
|

LOC4328196
Genomic Map