Assay Details
| Gene Symbol: LOC9266752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771439.2 | XP_015626925.1 | 18 - 19 | 3375 | 135 | |
| XM_015771440.2 | XP_015626926.1 | 18 - 19 | 3369 | 135 | ||
More Information
Set Membership: |
|

LOC9266752
Genomic Map