Assay Details
| Gene Symbol: LOC9266752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771439.2 | XP_015626925.1 | 1 - 2 | 316 | 111 | |
| XM_015771440.2 | XP_015626926.1 | 1 - 2 | 316 | 111 | ||
More Information
Set Membership: |
|

LOC9266752
Genomic Map