Assay Details
| Gene Symbol: LOC4328504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770832.2 | XP_015626318.1 | 1 - 2 | 163 | 68 | |
| XM_015770834.2 | XP_015626320.1 | 1 - 2 | 163 | 68 | ||
| GenBank mRNA | AK067511.1 | - | 1 - 2 | 171 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770832.2, XM_015770834.2 transcripts.
Set Membership: |
|

LOC4328504
Genomic Map