Assay Details
| Gene Symbol: LOC4328504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770832.2 | XP_015626318.1 | 7 - 8 | 743 | 89 | |
| XM_015770834.2 | XP_015626320.1 | 6 - 7 | 697 | 89 | ||
| GenBank mRNA | AK067511.1 | - | 5 - 6 | 2179 | 89 | |
More Information
Set Membership: |
|

LOC4328504
Genomic Map