Assay Details
| Gene Symbol: LOC4328504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770832.2 | XP_015626318.1 | 17 - 18 | 2783 | 132 | |
| XM_015770834.2 | XP_015626320.1 | 16 - 17 | 2737 | 132 | ||
| GenBank mRNA | AK067511.1 | - | 9 - 10 | 3247 | 132 | |
More Information
Set Membership: |
|

LOC4328504
Genomic Map