Assay Details
| Gene Symbol: LOC4328604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK120869.1 | - | 8 - 9 | 1841 | 93 | |
| AK121281.1 | - | 8 - 9 | 1809 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4328604| Gene Symbol: LOC4328604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK120869.1 | - | 8 - 9 | 1841 | 93 | |
| AK121281.1 | - | 8 - 9 | 1809 | 93 | ||
Set Membership: |
|