Assay Details
| Gene Symbol: LOC4329024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767185.1 | XP_015622671.1 | 1 - 2 | 163 | 78 | |
| XM_015767186.1 | XP_015622672.1 | 1 - 2 | 163 | 78 | ||
| XM_015767191.1 | XP_015622677.1 | 1 - 2 | 163 | 78 | ||
| XM_015767192.2 | XP_015622678.1 | 1 - 2 | 162 | 78 | ||
| GenBank mRNA | AK121583.1 | - | 1 - 2 | 74 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767185.1, XM_015767186.1, XM_015767191.1, XM_015767192.2 transcripts.
Set Membership: |
|

LOC4329024
Genomic Map