Assay Details
| Gene Symbol: LOC4329352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771680.2 | XP_015627166.1 | 10 - 11 | 2424 | 97 | |
| XM_015771682.2 | XP_015627168.1 | 10 - 11 | 2424 | 97 | ||
| GenBank mRNA | AK122152.1 | - | 3 - 4 | 1711 | 97 | |
More Information
Set Membership: |
|

LOC4329352
Genomic Map