Assay Details
| Gene Symbol: LOC4329532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772017.2 | XP_015627503.1 | 13 - 14 | 1838 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4329532| Gene Symbol: LOC4329532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772017.2 | XP_015627503.1 | 13 - 14 | 1838 | 93 | |
Set Membership: |
|