Assay Details
| Gene Symbol: LOC4329532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772017.2 | XP_015627503.1 | 14 - 15 | 1964 | 129 | |
| GenBank mRNA | EU735074.1 | - | 14 - 15 | 1588 | 129 | |
More Information
Set Membership: |
|

LOC4329532
Genomic Map