Assay Details
| Gene Symbol: LOC4329532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772017.2 | XP_015627503.1 | 19 - 20 | 2539 | 144 | |
| GenBank mRNA | AK108535.1 | - | 3 - 4 | 230 | 144 | |
| EU735074.1 | - | 19 - 20 | 2163 | 144 | ||
More Information
Set Membership: |
|

LOC4329532
Genomic Map