Assay Details
| Gene Symbol: LOC4329532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772017.2 | XP_015627503.1 | 21 - 22 | 2772 | 70 | |
| GenBank mRNA | AK108535.1 | - | 5 - 6 | 463 | 70 | |
| EU735074.1 | - | 21 - 22 | 2396 | 70 | ||
More Information
Set Membership: |
|

LOC4329532
Genomic Map