Assay Details
| Gene Symbol: LOC4329532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772017.2 | XP_015627503.1 | 9 - 10 | 1353 | 72 | |
| GenBank mRNA | EU735074.1 | - | 9 - 10 | 977 | 72 | |
More Information
Set Membership: |
|

LOC4329532
Genomic Map