Assay Details
| Gene Symbol: LOC4329561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769787.2 | XP_015625273.1 | 4 - 5 | 1275 | 96 | |
| GenBank mRNA | AB073749.1 | - | 4 - 5 | 1263 | 96 | |
| AK121403.1 | - | 4 - 5 | 1268 | 96 | ||
| AY578158.1 | - | 4 - 5 | 1218 | 96 | ||
More Information
Set Membership: |
|

LOC4329561
Genomic Map