Assay Details
| Gene Symbol: LOC4329561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769787.2 | XP_015625273.1 | 6 - 7 | 1623 | 61 | |
| GenBank mRNA | AB073749.1 | - | 6 - 7 | 1611 | 61 | |
| AK121403.1 | - | 6 - 7 | 1616 | 61 | ||
| AY578158.1 | - | 6 - 7 | 1566 | 61 | ||
More Information
Set Membership: |
|

LOC4329561
Genomic Map