Assay Details
| Gene Symbol: LOC4329582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767913.2 | XP_015623399.1 | 1 | 851 | 81 | |
| GenBank mRNA | AK060001.1 | - | 1 | 814 | 81 | |
| AK061396.1 | - | 1 | 817 | 81 | ||
| AK065946.1 | - | 1 | 786 | 81 | ||
| AK104564.1 | - | 1 | 785 | 81 | ||
| AK104600.1 | - | 1 | 817 | 81 | ||
| AK109400.2 | - | 1 | 817 | 81 | ||
| AK109402.2 | - | 1 | 838 | 81 | ||
More Information
Set Membership: |
|

LOC4329582
Genomic Map