Assay Details
| Gene Symbol: LOC4329648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770636.2 | XP_015626122.1 | 11 - 12 | 1317 | 93 | |
| GenBank mRNA | AK060250.1 | - | 11 - 12 | 1319 | 93 | |
| AK069920.1 | - | 11 - 12 | 1320 | 93 | ||
More Information
Set Membership: |
|

LOC4329648
Genomic Map