Assay Details
| Gene Symbol: LOC4329648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770636.2 | XP_015626122.1 | 1 - 2 | 156 | 76 | |
| GenBank mRNA | AK060250.1 | - | 1 - 2 | 158 | 76 | |
| AK069920.1 | - | 1 - 2 | 159 | 76 | ||
More Information
Set Membership: |
|

LOC4329648
Genomic Map