Assay Details
| Gene Symbol: LOC4329657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772155.1 | XP_015627641.1 | 1 - 2 | 381 | 85 | |
| GenBank mRNA | AK108057.1 | - | 1 - 2 | 381 | 85 | |
More Information
Set Membership: |
|

LOC4329657
Genomic Map