Assay Details
| Gene Symbol: LOC4329757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772010.2 | XP_015627496.1 | 1 - 2 | 284 | 90 | |
| GenBank mRNA | AK059578.1 | - | 1 - 2 | 278 | 90 | |
| AK102294.1 | - | 1 - 2 | 288 | 90 | ||
More Information
Set Membership: |
|

LOC4329757
Genomic Map