Assay Details
| Gene Symbol: LOC4329760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771416.2 | XP_015626902.1 | 2 - 3 | 1005 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4329760| Gene Symbol: LOC4329760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771416.2 | XP_015626902.1 | 2 - 3 | 1005 | 134 | |
Set Membership: |
|