Assay Details
| Gene Symbol: LOC4329760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771416.2 | XP_015626902.1 | 6 - 7 | 1697 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4329760| Gene Symbol: LOC4329760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771416.2 | XP_015626902.1 | 6 - 7 | 1697 | 116 | |
Set Membership: |
|