Assay Details
| Gene Symbol: LOC4329868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771761.2 | XP_015627247.1 | 3 - 4 | 761 | 126 | |
| XM_015771762.2 | XP_015627248.1 | 2 - 3 | 1716 | 126 | ||
| XM_015771763.2 | XP_015627249.1 | 3 - 4 | 963 | 126 | ||
| GenBank mRNA | AK102225.1 | - | 3 - 4 | 753 | 126 | |
More Information
Set Membership: |
|

LOC4329868
Genomic Map