Assay Details
Gene Symbol: LOC4329911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770238.2 | XP_015625724.1 | 6 - 7 | 842 | 103 | |
GenBank mRNA | AK105762.1 | - | 6 - 7 | 817 | 103 | |
AK120769.1 | - | 6 - 7 | 810 | 103 | ||
AY224457.1 | - | 3 - 4 | 248 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
