Assay Details
| Gene Symbol: LOC4330240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772207.2 | XP_015627693.1 | 1 - 2 | 295 | 96 | |
| GenBank mRNA | AK068162.1 | - | 1 - 2 | 259 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772207.2 transcripts.
Set Membership: |
|

LOC4330240
Genomic Map