Assay Details
| Gene Symbol: LOC4330240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772207.2 | XP_015627693.1 | 2 - 3 | 1493 | 79 | |
| XM_015772208.2 | XP_015627694.1 | 2 - 3 | 1496 | 79 | ||
| GenBank mRNA | AK068162.1 | - | 2 - 3 | 1457 | 79 | |
More Information
Set Membership: |
|

LOC4330240
Genomic Map