Assay Details
| Gene Symbol: LOC4330259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771376.1 | XP_015626862.1 | 2 - 3 | 2226 | 64 | |
| GenBank mRNA | AK101871.1 | - | 2 - 3 | 2234 | 64 | |
More Information
Set Membership: |
|

LOC4330259
Genomic Map