Assay Details
| Gene Symbol: LOC4330259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771376.1 | XP_015626862.1 | 4 - 5 | 2725 | 106 | |
| GenBank mRNA | AK101871.1 | - | 4 - 5 | 2733 | 106 | |
More Information
Set Membership: |
|

LOC4330259
Genomic Map