Assay Details
| Gene Symbol: LOC4330264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768064.2 | XP_015623550.1 | 4 - 5 | 967 | 91 | |
| GenBank mRNA | AK059531.1 | - | 4 - 5 | 901 | 91 | |
| AK103585.1 | - | 4 - 5 | 883 | 91 | ||
More Information
Set Membership: |
|

LOC4330264
Genomic Map