Assay Details
| Gene Symbol: LOC4330561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771775.2 | XP_015627261.1 | 1 - 2 | 283 | 77 | |
| XM_015771776.2 | XP_015627262.1 | 1 - 2 | 282 | 77 | ||
| XM_015771777.2 | XP_015627263.1 | 1 - 2 | 283 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771775.2, XM_015771776.2, XM_015771777.2 transcripts.
Set Membership: |
|

LOC4330561
Genomic Map