Assay Details
| Gene Symbol: LOC4330868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772116.2 | XP_015627602.1 | 11 - 12 | 2034 | 103 | |
| GenBank mRNA | AB040053.1 | - | 11 - 12 | 1851 | 103 | |
| AK102740.1 | - | 11 - 12 | 2041 | 103 | ||
| AK111614.1 | - | 11 - 12 | 1860 | 103 | ||
| AK112098.1 | - | 12 - 13 | 1558 | 103 | ||
More Information
Set Membership: |
|

LOC4330868
Genomic Map