Assay Details
| Gene Symbol: LOC4330868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772116.2 | XP_015627602.1 | 5 - 6 | 1349 | 67 | |
| GenBank mRNA | AB040053.1 | - | 5 - 6 | 1166 | 67 | |
| AK111614.1 | - | 5 - 6 | 1175 | 67 | ||
| AK112098.1 | - | 6 - 7 | 873 | 67 | ||
More Information
Set Membership: |
|

LOC4330868
Genomic Map