Assay Details
| Gene Symbol: LOC4330868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772116.2 | XP_015627602.1 | 8 - 9 | 1648 | 101 | |
| GenBank mRNA | AB040053.1 | - | 8 - 9 | 1465 | 101 | |
| AK102740.1 | - | 8 - 9 | 1655 | 101 | ||
| AK111614.1 | - | 8 - 9 | 1474 | 101 | ||
More Information
Set Membership: |
|

LOC4330868
Genomic Map