Assay Details
| Gene Symbol: LOC4330921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768091.2 | XP_015623577.1 | 8 - 9 | 1041 | 119 | |
| GenBank mRNA | AK067608.1 | - | 8 - 9 | 999 | 119 | |
| AK121526.1 | - | 8 - 9 | 1042 | 119 | ||
More Information
Set Membership: |
|

LOC4330921
Genomic Map