Assay Details
| Gene Symbol: LOC4330978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772169.2 | XP_015627655.1 | 4 - 5 | 1266 | 146 | |
| GenBank mRNA | AK066282.1 | - | 4 - 5 | 363 | 146 | |
| AK099697.1 | - | 4 - 5 | 1271 | 146 | ||
| AY445625.1 | - | 4 - 5 | 344 | 146 | ||
More Information
Set Membership: |
|

LOC4330978
Genomic Map