Assay Details
| Gene Symbol: LOC4330978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772169.2 | XP_015627655.1 | 8 - 9 | 1738 | 112 | |
| GenBank mRNA | AK066282.1 | - | 8 - 9 | 835 | 112 | |
| AK099697.1 | - | 8 - 9 | 1743 | 112 | ||
| AY445625.1 | - | 8 - 9 | 816 | 112 | ||
More Information
Set Membership: |
|

LOC4330978
Genomic Map