Assay Details
| Gene Symbol: LOC4330987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770018.2 | XP_015625504.1 | 18 - 19 | 3428 | 83 | |
| GenBank mRNA | AK072317.1 | - | 16 - 17 | 1912 | 83 | |
| BT132117.1 | - | 1 - 2 | 47 | 83 | ||
More Information
Set Membership: |
|

LOC4330987
Genomic Map