Assay Details
| Gene Symbol: LOC4330987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770018.2 | XP_015625504.1 | 19 - 20 | 3519 | 87 | |
| GenBank mRNA | AK072317.1 | - | 17 - 18 | 2003 | 87 | |
| BT132117.1 | - | 2 - 3 | 138 | 87 | ||
More Information
Set Membership: |
|

LOC4330987
Genomic Map