Assay Details
| Gene Symbol: LOC4330987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770018.2 | XP_015625504.1 | 7 - 8 | 2140 | 78 | |
| GenBank mRNA | AK072317.1 | - | 5 - 6 | 624 | 78 | |
More Information
Set Membership: |
|

LOC4330987
Genomic Map