Assay Details
| Gene Symbol: LOC4331022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770157.2 | XP_015625643.1 | 4 - 5 | 3420 | 77 | |
| XM_015770158.2 | XP_015625644.1 | 4 - 5 | 3420 | 77 | ||
| GenBank mRNA | AK061459.1 | - | 2 - 3 | 626 | 77 | |
| AK104480.1 | - | 2 - 3 | 624 | 77 | ||
More Information
Set Membership: |
|

LOC4331022
Genomic Map