Assay Details
Gene Symbol: LOC4331069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770166.1 | XP_015625652.1 | 4 - 5 | 554 | 134 | |
GenBank mRNA | AK073740.1 | - | 4 - 5 | 560 | 134 |
Gene Symbol: LOC4340405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK065642.1 | - | 142 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
