Assay Details
Gene Symbol: LOC4331069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770166.1 | XP_015625652.1 | 6 - 7 | 812 | 80 | |
GenBank mRNA | AK073740.1 | - | 6 - 7 | 818 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
