Assay Details
| Gene Symbol: LOC4331193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770085.2 | XP_015625571.1 | 5 - 6 | 831 | 104 | |
| GenBank mRNA | AK122065.1 | - | 5 - 6 | 786 | 104 | |
More Information
Set Membership: |
|

LOC4331193
Genomic Map